siRNA Details
| Gene Symbol: C20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009984.2 | NP_001009984.1 | 21 | 1862 | ||
| XM_005260684.3 | XP_005260741.1 | 21 | 1856 | |||
| XM_005260687.4 | XP_005260744.1 | 2 | 698 | |||
| XM_011529208.2 | XP_011527510.1 | 2 | 199 | |||
| XM_017027783.1 | XP_016883272.1 | 18 | 1483 | |||
| GenBank mRNA | AK023919.1 | - | 3 | 524 | ||
| AK226100.1 | - | 16 | 1286 | |||
| AK303375.1 | - | 12 | 1231 | |||
| AL110249.1 | - | 12 | 827 | |||
| BC122529.1 | - | 12 | 1080 | 40012183 | ||

C20orf194
Genomic Map