siRNA Details
Gene Symbol: C2orf27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013310.3 | NP_037442.3 | 1 | 43 | ||
GenBank mRNA | AF038169.1 | - | 1 | 43 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C2orf27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013310.3 | NP_037442.3 | 1 | 43 | ||
GenBank mRNA | AF038169.1 | - | 1 | 43 |