siRNA Details
| Gene Symbol: CCDC137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199287.2 | NP_954981.1 | 6 | 1217 | ||
| XM_011524738.2 | XP_011523040.1 | 6 | 1119 | |||
| GenBank mRNA | FJ649645.1 | - | 6 | 2907 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC137| Gene Symbol: CCDC137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199287.2 | NP_954981.1 | 6 | 1217 | ||
| XM_011524738.2 | XP_011523040.1 | 6 | 1119 | |||
| GenBank mRNA | FJ649645.1 | - | 6 | 2907 | ||