siRNA Details
| Gene Symbol: PABPC1L2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012977.2 | NP_001012995.1 | 1 | 2026 | ||
| GenBank mRNA | AY730282.1 | - | 1 | 425 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PABPC1L2A| Gene Symbol: PABPC1L2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012977.2 | NP_001012995.1 | 1 | 2026 | ||
| GenBank mRNA | AY730282.1 | - | 1 | 425 | ||