siRNA Details
| Gene Symbol: TEX36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128202.2 | NP_001121674.1 | 1 | 89 | ||
| NM_001318133.1 | NP_001305062.1 | 1 | 89 | |||
| XM_005269817.4 | XP_005269874.1 | 1 | 87 | |||
| GenBank mRNA | BC062717.1 | - | 1 | 92 | 6101709 | |
| BX336391.2 | - | 1 | 51 | |||

TEX36
Genomic Map