siRNA Details
| Gene Symbol: ZC2HC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013623.2 | NP_001013645.1 | 3 | 230 | ||
| GenBank mRNA | BC146950.1 | - | 3 | 230 | 9056878 | |
| BC146969.1 | - | 3 | 230 | 9056897 | ||
| BC171862.1 | - | 4 | 313 | 9054516 | ||

ZC2HC1B
Genomic Map