siRNA Details
| Gene Symbol: CNKSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173515.2 | NP_775786.2 | 6 | 1144 | ||
| XM_011535485.2 | XP_011533787.1 | 6 | 3627 | |||
| GenBank mRNA | AK055911.1 | - | 6 | 1184 | ||
| AK309680.1 | - | 6 | 750 | |||
| AY328891.1 | - | 6 | 1144 | |||
| AY328892.1 | - | 6 | 1144 | |||
| AY328893.1 | - | 6 | 1144 | |||
| AY328894.1 | - | 3 | 427 | |||
| BC060761.1 | - | 6 | 1066 | 5299236 | ||

CNKSR3
Genomic Map