siRNA Details
| Gene Symbol: EIF4EBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004096.4 | NP_004087.1 | 1 | 389 | ||
| GenBank mRNA | AK313724.1 | - | 1 | 349 | ||
| BC005057.1 | - | 1 | 115 | 2823322 | ||
| BC050633.1 | - | 1 | 328 | 5555034 | ||
| BG717474.1 | - | 1 | 393 | |||
| BT007317.1 | - | 1 | 92 | |||
| CR542240.1 | - | 1 | 92 | |||
| L36056.1 | - | 1 | 92 | |||

EIF4EBP2
Genomic Map