siRNA Details
| Gene Symbol: SMIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001124767.1 | NP_001118239.1 | 2 | 424 | ||
| GenBank mRNA | AK095910.1 | - | 2 | 1065 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM4| Gene Symbol: SMIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001124767.1 | NP_001118239.1 | 2 | 424 | ||
| GenBank mRNA | AK095910.1 | - | 2 | 1065 | ||