siRNA Details
| Gene Symbol: C9orf129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098808.1 | NP_001092278.1 | 5 | 1016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf129| Gene Symbol: C9orf129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098808.1 | NP_001092278.1 | 5 | 1016 | ||