siRNA Details
| Gene Symbol: RBMXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162536.2 | NP_001156008.1 | 3 | 3613 | ||
| NM_019610.5 | NP_062556.2 | 2 | 3513 | |||
| GenBank mRNA | AL832554.1 | - | 2 | 3366 | ||
| AL832785.1 | - | 1 | 2454 | |||
| BC012942.1 | - | 2 | 3172 | 3875565 | ||

RBMXL1
Genomic Map