siRNA Details
| Gene Symbol: FBXL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099784.2 | NP_001093254.2 | 11 | 3765 | ||
| NM_001282351.1 | NP_001269280.1 | 11 | 3633 | |||
| GenBank mRNA | AK098777.1 | - | 10 | 1657 | ||
| AK127701.1 | - | 11 | 3617 | |||
| AL137589.1 | - | 3 | 1975 | |||
| BC059173.1 | - | 10 | 3281 | 5738074 | ||

FBXL19
Genomic Map