siRNA Details
| Gene Symbol: ATXN7L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098833.1 | NP_001092303.1 | 12 | 1835 | ||
| NM_020218.1 | NP_064603.1 | 12 | 1856 | |||
| XM_011525038.2 | XP_011523340.1 | 12 | 2507 | |||
| XM_011525044.2 | XP_011523346.1 | 12 | 2450 | |||
| XM_011525046.2 | XP_011523348.1 | 12 | 2429 | |||
| XM_017024880.1 | XP_016880369.1 | 13 | 2085 | |||
| XM_017024881.1 | XP_016880370.1 | 13 | 2384 | |||
| XM_017024882.1 | XP_016880371.1 | 12 | 2486 | |||
| XM_017024883.1 | XP_016880372.1 | 13 | 2088 | |||
| XM_017024884.1 | XP_016880373.1 | 12 | 2110 | |||
| XM_017024885.1 | XP_016880374.1 | 13 | 2031 | |||
| XM_017024886.1 | XP_016880375.1 | 13 | 2010 | |||
| GenBank mRNA | AK056002.1 | - | 9 | 1379 | ||
| AK074344.1 | - | 1 | 444 | |||
| AL390158.1 | - | 7 | 1264 | |||
| BC024695.2 | - | 5 | 1193 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001098836, NM_001098837, AK053165, BC130269, BC059880, and others.

ATXN7L3
Genomic Map