siRNA Details
| Gene Symbol: HIGD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109619.1 | NP_001103089.1 | 3 | 260 | ||
| XM_017019784.1 | XP_016875273.1 | 6 | 1835 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIGD1C| Gene Symbol: HIGD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109619.1 | NP_001103089.1 | 3 | 260 | ||
| XM_017019784.1 | XP_016875273.1 | 6 | 1835 | |||