siRNA Details
| Gene Symbol: KRTAP2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033032.2 | NP_149021.2 | 1 | 695 | ||
| GenBank mRNA | AJ302536.1 | - | 582 | |||
| Gene Symbol: KRTAP2-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ406929.1 | - | 516 | |||

KRTAP2-2
Genomic Map