siRNA Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109977.1 | NP_001103447.1 | 4 | 650 | ||
| XM_005263199.1 | XP_005263256.1 | 3 | 525 | |||
| XM_011532220.1 | XP_011530522.1 | 2 | 403 | |||
| XM_011532221.2 | XP_011530523.1 | 4 | 723 | |||
| XM_011532222.2 | XP_011530524.1 | 3 | 645 | |||
| XM_011532223.2 | XP_011530525.1 | 2 | 520 | |||
| XM_011532224.2 | XP_011530526.1 | 2 | 219 | |||
| GenBank mRNA | AK125363.1 | - | 4 | 513 | ||
| BC014937.1 | - | 2 | 234 | |||
| DA722299.1 | - | 3 | 525 | |||

FAM160A1
Genomic Map