siRNA Details
| Gene Symbol: GGACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195087.1 | NP_001182016.1 | 3 | 951 | ||
| NM_033110.2 | NP_149101.1 | 2 | 966 | |||
| XM_005254083.2 | XP_005254140.1 | 2 | 1093 | |||
| XM_011521129.2 | XP_011519431.1 | 3 | 3929 | |||
| XM_017020804.1 | XP_016876293.1 | 3 | 1067 | |||
| GenBank mRNA | AK091974.1 | - | 3 | 639 | ||
| AK093228.1 | - | 1 | 182 | |||
| AK095850.1 | - | 3 | 951 | |||
| BC001077.2 | - | 2 | 907 | |||
| BC004360.1 | - | 2 | 907 | |||

GGACT
Genomic Map