siRNA Details
Gene Symbol: CCDC144B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036647.1 | 9 | 2150 | |||
GenBank mRNA | BC101585.1 | - | 9 | 2065 | 8069091 | |
BC104815.1 | - | 9 | 2065 | 8143818 | ||
BC143561.1 | - | 9 | 2062 | 9052072 | ||
BX647826.1 | - | 9 | 2141 |
Gene Symbol: CCDC144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014695.2 | NP_055510.1 | 9 | 2056 | ||
NR_130142.1 | 9 | 2056 | ||||
XM_017025429.1 | XP_016880918.1 | 9 | 2073 | |||
XM_017025430.1 | XP_016880919.1 | 9 | 2146 | |||
GenBank mRNA | AK125971.1 | - | 5 | 431 | ||
BC133019.1 | - | 9 | 2056 | 40147041 | ||
BC144224.1 | - | 9 | 2056 | 9052744 | ||
BC144225.1 | - | 9 | 2056 | 9052745 |
Gene Symbol: CCDC144CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023380.1 | 9 | 2127 | |||
GenBank mRNA | DV080283.1 | - | 2 | 151 |
