siRNA Details
| Gene Symbol: KHDC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017361.2 | NP_001017361.1 | 3 | 460 | ||
| GenBank mRNA | AB211062.1 | - | 3 | 460 | ||
| BC132844.1 | - | 3 | 452 | 40146866 | ||
| BC137160.1 | - | 3 | 452 | 9021157 | ||

KHDC3L
Genomic Map