siRNA Details
Gene Symbol: CCDC167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138493.2 | NP_612502.1 | 4 | 415 | ||
GenBank mRNA | AF161383.1 | - | 4 | 414 | ||
BC003515.1 | - | 4 | 360 | |||
BC108655.1 | - | 4 | 388 | 6053765 | ||
BE907609.1 | - | 3 | 300 |
