siRNA Details
| Gene Symbol: CCDC167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138493.2 | NP_612502.1 | 3 | 228 | ||
| GenBank mRNA | AF161383.1 | - | 3 | 227 | ||
| BC003515.1 | - | 3 | 173 | |||
| BC104835.1 | - | 3 | 224 | 8143838 | ||
| BC108655.1 | - | 3 | 201 | 6053765 | ||
| BE907609.1 | - | 2 | 113 | |||
| DA152306.1 | - | 3 | 228 | |||

CCDC167
Genomic Map