siRNA Details
| Gene Symbol: AMOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113490.1 | NP_001106962.1 | 10 | 3120 | ||
| NM_133265.2 | NP_573572.1 | 11 | 2689 | |||
| XM_005262087.1 | XP_005262144.1 | 13 | 3865 | |||
| XM_005262090.1 | XP_005262147.1 | 10 | 2362 | |||
| XM_011530875.2 | XP_011529177.1 | 14 | 3977 | |||
| XM_017029289.1 | XP_016884778.1 | 12 | 2893 | |||
| GenBank mRNA | AB028994.3 | - | 9 | 1893 | ||
| AF286598.1 | - | 11 | 2689 | |||
| AY987378.1 | - | 10 | 3120 | |||
| BC130294.1 | - | 9 | 2113 | 40146325 | ||

AMOT
Genomic Map