siRNA Details
| Gene Symbol: CRYGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144727.2 | NP_653328.1 | 2, 3 | 526 | ||
| XM_005249952.3 | XP_005250009.1 | 2, 3 | 526 | |||
| XM_011515836.1 | XP_011514138.1 | 2, 3 | 526 | |||
| XM_011515837.2 | XP_011514139.1 | 2, 3 | 905 | |||
| GenBank mRNA | AF445455.1 | - | 2, 3 | 369 | ||
| BC100878.2 | - | 2, 3 | 364 | 40003204 | ||
| BC100880.2 | - | 2, 3 | 364 | 40003208 | ||
| BC100881.2 | - | 2, 3 | 364 | 40003209 | ||

CRYGN
Genomic Map