siRNA Details
| Gene Symbol: ZNF783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195220.1 | NP_001182149.1 | 6 | 1000 | ||
| XM_011515701.2 | XP_011514003.1 | 6 | 928 | |||
| GenBank mRNA | BG029497.1 | - | 5 | 572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF783| Gene Symbol: ZNF783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195220.1 | NP_001182149.1 | 6 | 1000 | ||
| XM_011515701.2 | XP_011514003.1 | 6 | 928 | |||
| GenBank mRNA | BG029497.1 | - | 5 | 572 | ||