siRNA Details
| Gene Symbol: ZNF746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152557.4 | NP_689770.3 | 6, 7 | 1113 | ||
| XM_005249956.2 | XP_005250013.1 | 6, 7 | 1183 | |||
| GenBank mRNA | AK055975.1 | - | 1 | 12 | ||
| AK128244.1 | - | 4, 5 | 473 | |||
| BC068505.1 | - | 6, 7 | 1009 | 30347892 | ||

ZNF746
Genomic Map