siRNA Details
| Gene Symbol: SLC2A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207420.2 | NP_997303.2 | 3 | 202 | ||
| XM_011540824.2 | XP_011539126.1 | 3 | 241 | |||
| XM_011540825.2 | XP_011539127.1 | 3 | 241 | |||
| GenBank mRNA | AY571960.2 | - | 3 | 202 | ||

SLC2A7
Genomic Map