siRNA Details
| Gene Symbol: WBSCR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152559.2 | NP_689772.2 | 5 | 488 | ||
| XM_017011777.1 | XP_016867266.1 | 5 | 490 | |||
| XM_017011778.1 | XP_016867267.1 | 5 | 612 | |||
| GenBank mRNA | AF534110.1 | - | 5 | 499 | ||
| AK301597.1 | - | 4 | 343 | |||
| AY354928.1 | - | 5 | 498 | |||
| BC030295.2 | - | 4 | 452 | 5396423 | ||

WBSCR27
Genomic Map