siRNA Details
| Gene Symbol: RBM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053043.2 | NP_444271.2 | 17, 18 | 3837 | ||
| XM_005249531.1 | XP_005249588.1 | 18, 19 | 4005 | |||
| XM_005249532.1 | XP_005249589.1 | 18, 19 | 4002 | |||
| XM_005249533.1 | XP_005249590.1 | 18, 19 | 3975 | |||
| XM_005249534.1 | XP_005249591.1 | 17, 18 | 3885 | |||
| XM_005249535.1 | XP_005249592.1 | 17, 18 | 3855 | |||
| XM_011515840.1 | XP_011514142.1 | 17, 18 | 3795 | |||
| GenBank mRNA | AK056415.1 | - | 2, 3 | 245 | ||
| AL832196.1 | - | 7, 8 | 571 | |||
| BC093947.1 | - | 3, 4 | 343 | |||
| BC093949.1 | - | 3, 4 | 343 | |||

RBM33
Genomic Map