siRNA Details
| Gene Symbol: AGR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176813.4 | NP_789783.1 | 5, 6 | 371 | ||
| XM_005249627.2 | XP_005249684.1 | 5, 6 | 350 | |||
| XM_011515152.2 | XP_011513454.1 | 5, 6 | 363 | |||
| GenBank mRNA | AY069977.1 | - | 4, 5 | 336 | ||
| AY359065.1 | - | 5, 6 | 355 | |||
| BC058284.1 | - | 5, 6 | 376 | 4694757 | ||
| BG540617.1 | - | 373 | ||||

AGR3
Genomic Map