siRNA Details
| Gene Symbol: CDCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317906.1 | NP_001304835.1 | 9 | 1862 | ||
| NM_001317907.1 | NP_001304836.1 | 10 | 1779 | |||
| NM_152562.3 | NP_689775.2 | 10 | 1779 | |||
| XM_006716294.2 | XP_006716357.1 | 10 | 1794 | |||
| XM_011544415.1 | XP_011542717.1 | 11 | 1866 | |||
| XM_011544416.2 | XP_011542718.1 | 11 | 1560 | |||
| XM_011544417.2 | XP_011542719.1 | 11 | 1617 | |||
| XM_011544418.1 | XP_011542720.1 | 11 | 1830 | |||
| GenBank mRNA | AK092164.1 | - | 5 | 396 | ||
| AK292505.1 | - | 9 | 1594 | |||
| AK299278.1 | - | 10 | 1485 | |||
| AL833396.1 | - | 7 | 884 | |||
| AL833627.1 | - | 9 | 1556 | |||
| BC036214.1 | - | 7 | 907 | 5267276 | ||
| BX649008.1 | - | 6 | 770 | |||

CDCA2
Genomic Map