siRNA Details
| Gene Symbol: SBSPON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153225.3 | NP_694957.3 | 2, 3 | 618 | ||
| XM_017013145.1 | XP_016868634.1 | 2, 3 | 863 | |||
| GenBank mRNA | AK292970.1 | - | 2, 3 | 518 | ||
| AY040546.1 | - | 2, 3 | 516 | |||
| BC042877.1 | - | 2, 3 | 618 | 5169941 | ||

SBSPON
Genomic Map