siRNA Details
| Gene Symbol: CAMSAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015447.3 | NP_056262.3 | 16 | 4472 | ||
| XM_005263396.3 | XP_005263453.1 | 17 | 4897 | |||
| XM_005263397.1 | XP_005263454.1 | 14 | 3998 | |||
| XM_011518272.1 | XP_011516574.1 | 16 | 4166 | |||
| XM_011518273.2 | XP_011516575.1 | 15 | 3997 | |||
| XM_011518274.1 | XP_011516576.1 | 14 | 4027 | |||
| XM_017014301.1 | XP_016869790.1 | 16 | 4753 | |||
| XM_017014302.1 | XP_016869791.1 | 16 | 4677 | |||
| GenBank mRNA | AJ519841.1 | - | 14 | 3997 | ||
| AL834528.1 | - | 6 | 824 | |||
| BC010646.1 | - | 6 | 1198 | |||
| BC012778.2 | - | 6 | 2472 | |||
| BC130580.1 | - | 16 | 4131 | 40146611 | ||
| BC144082.1 | - | 16 | 4131 | 9052602 | ||

CAMSAP1
Genomic Map