siRNA Details
| Gene Symbol: C1QTNF9B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014442.2 | NP_001014442.2 | 4 | 786 | ||
| NM_001135816.1 | NP_001129288.1 | 4 | 683 | |||
| GenBank mRNA | BC015452.1 | - | 4 | 788 | ||
| BC073902.1 | - | 4 | 786 | |||
| BC142999.1 | - | 4 | 683 | |||

C1QTNF9B-AS1
Genomic Map