siRNA Details
| Gene Symbol: CAMSAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015447.3 | NP_056262.3 | 17 | 4591 | ||
| XM_005263396.3 | XP_005263453.1 | 18 | 5016 | |||
| XM_005263397.1 | XP_005263454.1 | 15 | 4117 | |||
| XM_011518272.1 | XP_011516574.1 | 17 | 4285 | |||
| XM_011518273.2 | XP_011516575.1 | 16 | 4116 | |||
| XM_011518274.1 | XP_011516576.1 | 15 | 4146 | |||
| XM_017014301.1 | XP_016869790.1 | 17 | 4872 | |||
| XM_017014302.1 | XP_016869791.1 | 17 | 4796 | |||
| GenBank mRNA | AJ519841.1 | - | 15 | 4116 | ||
| AL834528.1 | - | 7 | 943 | |||
| BC010646.1 | - | 7 | 1317 | |||
| BC012778.2 | - | 7 | 2591 | |||
| BC130580.1 | - | 17 | 4250 | 40146611 | ||
| BC144082.1 | - | 17 | 4250 | 9052602 | ||

CAMSAP1
Genomic Map