siRNA Details
| Gene Symbol: MAMDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206920.2 | NP_996803.2 | 9, 10 | 1080 | ||
| GenBank mRNA | AY358419.1 | - | 9, 10 | 1080 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAMDC4| Gene Symbol: MAMDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206920.2 | NP_996803.2 | 9, 10 | 1080 | ||
| GenBank mRNA | AY358419.1 | - | 9, 10 | 1080 | ||