siRNA Details
| Gene Symbol: LCN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198946.2 | NP_945184.1 | 3 | 302 | ||
| GenBank mRNA | AF303084.1 | - | 3 | 299 | ||
| AY301269.1 | - | 3 | 257 | |||
| AY358598.1 | - | 3 | 282 | |||
| BC062746.1 | - | 3 | 303 | 6452008 | ||

LCN6
Genomic Map