siRNA Details
| Gene Symbol: OR1Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012364.1 | NP_036496.1 | 1 | 823 | ||
| GenBank mRNA | BC140713.1 | - | 1 | 892 | 9021581 | |
| X89667.1 | - | 1 | 449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR1Q1| Gene Symbol: OR1Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012364.1 | NP_036496.1 | 1 | 823 | ||
| GenBank mRNA | BC140713.1 | - | 1 | 892 | 9021581 | |
| X89667.1 | - | 1 | 449 | |||