siRNA Details
| Gene Symbol: FAM201A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027294.1 | 2 | 988 | |||
| GenBank mRNA | BC036230.1 | - | 2 | 987 | ||
| BC041970.1 | - | 2 | 999 | |||
| Gene Symbol: LOC101929748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136301.1 | 1 | 808 | |||
| GenBank mRNA | BC040975.2 | - | 1 | 233 | ||

FAM201A
Genomic Map