siRNA Details
Gene Symbol: ATP8B5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003582.1 | 11 | 1626 | |||
NR_110320.1 | 11 | 1622 | ||||
GenBank mRNA | BC035229.1 | - | 10 | 3032 | ||
BC130448.1 | - | 6 | 1386 | |||
BC130450.1 | - | 6 | 1233 |
