siRNA Details
| Gene Symbol: FMR1NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152578.2 | NP_689791.1 | 5 | 796 | ||
| GenBank mRNA | AK098602.1 | - | 5 | 781 | ||
| AY211917.1 | - | 5 | 789 | |||
| BC034320.1 | - | 5 | 796 | 4836772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FMR1NB| Gene Symbol: FMR1NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152578.2 | NP_689791.1 | 5 | 796 | ||
| GenBank mRNA | AK098602.1 | - | 5 | 781 | ||
| AY211917.1 | - | 5 | 789 | |||
| BC034320.1 | - | 5 | 796 | 4836772 | ||