siRNA Details
| Gene Symbol: ARHGAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282607.1 | NP_001269536.1 | 2 | 254 | ||
| NM_144967.3 | NP_659404.2 | 2 | 490 | |||
| GenBank mRNA | AK128203.1 | - | 2 | 490 | ||
| AK294274.1 | - | 2 | 213 | |||
| AK296748.1 | - | 2 | 232 | |||
| BC063790.1 | - | 2 | 89 | 6196091 | ||

ARHGAP36
Genomic Map