siRNA Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031745.4 | NP_001026915.1 | 5 | 456 | ||
| NM_144968.3 | NP_659405.1 | 5 | 456 | |||
| XM_005261988.3 | XP_005262045.1 | 5 | 664 | |||
| GenBank mRNA | AK057345.1 | - | 5 | 422 | ||
| AK290346.1 | - | 5 | 406 | |||
| AK292871.1 | - | 5 | 411 | |||
| BC036767.1 | - | 5 | 371 | 5744127 | ||

RIBC1
Genomic Map