siRNA Details
| Gene Symbol: RHOXF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139282.2 | NP_644811.1 | 2 | 485 | ||
| XM_011531281.2 | XP_011529583.1 | 3 | 536 | |||
| GenBank mRNA | AY099086.1 | - | 2 | 485 | ||
| BC069324.1 | - | 2 | 460 | 7262157 | ||
| BC069529.1 | - | 2 | 462 | 7262169 | ||
| BC103857.1 | - | 2 | 444 | 40003132 | ||
| BC103858.1 | - | 2 | 444 | 40003136 | ||

RHOXF1
Genomic Map