siRNA Details
| Gene Symbol: CXorf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025265.2 | NP_001020436.1 | 4 | 498 | ||
| NR_033212.1 | 4 | 479 | ||||
| GenBank mRNA | BC144434.1 | - | 4 | 296 | ||
| DB457782.1 | - | 4 | 454 | |||
| DB523273.1 | - | 3 | 182 | |||

CXorf65
Genomic Map