siRNA Details
| Gene Symbol: AWAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013579.2 | NP_001013597.1 | 6 | 728 | ||
| XM_017029295.1 | XP_016884784.1 | 6 | 1894 | |||
| GenBank mRNA | AY947638.1 | - | 6 | 687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AWAT1| Gene Symbol: AWAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013579.2 | NP_001013597.1 | 6 | 728 | ||
| XM_017029295.1 | XP_016884784.1 | 6 | 1894 | |||
| GenBank mRNA | AY947638.1 | - | 6 | 687 | ||