siRNA Details
| Gene Symbol: USP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201286.3 | NP_958443.1 | 2 | 1394 | ||
| XM_017029299.1 | XP_016884788.1 | 3 | 1661 | |||
| XM_017029300.1 | XP_016884789.1 | 3 | 1544 | |||
| XM_017029301.1 | XP_016884790.1 | 2 | 1464 | |||
| GenBank mRNA | AJ583823.2 | - | 1 | 1315 | ||

USP51
Genomic Map