siRNA Details
| Gene Symbol: H2BFWT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002916.4 | NP_001002916.3 | 3 | 832 | ||
| GenBank mRNA | BC038109.2 | - | 3 | 829 | 5266336 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2BFWT| Gene Symbol: H2BFWT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002916.4 | NP_001002916.3 | 3 | 832 | ||
| GenBank mRNA | BC038109.2 | - | 3 | 829 | 5266336 | |