siRNA Details
| Gene Symbol: H2BFWT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002916.4 | NP_001002916.3 | 1 | 187 | ||
| GenBank mRNA | AY283370.1 | - | 1 | 160 | ||
| BC038109.2 | - | 1 | 184 | 5266336 | ||
| BC118604.1 | - | 1 | 180 | 40109007 | ||
| BC121816.1 | - | 1 | 180 | 40109006 | ||

H2BFWT
Genomic Map