siRNA Details
| Gene Symbol: C11orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173525.2 | NP_775796.2 | 2 | 681 | ||
| XM_011519926.2 | XP_011518228.1 | 2 | 3252 | |||
| XM_011519927.2 | XP_011518229.1 | 2 | 3252 | |||
| GenBank mRNA | BC031612.1 | - | 2 | 681 | 5171107 | |

C11orf42
Genomic Map