siRNA Details
Gene Symbol: ALDH1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034173.3 | NP_001029345.2 | 15 | 1969 | ||
NR_027752.1 | 15 | 1965 | ||||
XM_011537986.2 | XP_011536288.1 | 13 | 1528 | |||
XM_011537988.2 | XP_011536290.1 | 15 | 1990 | |||
XM_017018889.1 | XP_016874378.1 | 16 | 2175 | |||
GenBank mRNA | AK095827.1 | - | 15 | 1864 | ||
AK300373.1 | - | 15 | 1864 | |||
BC103934.1 | - | 15 | 1864 | 40008174 | ||
BC103935.1 | - | 15 | 1860 | 40008176 | ||
CR627287.1 | - | 11 | 1833 |

More Information
Panther Classification:
Molecular Function -




Gene Ontology Categories:
